Water purification system, PURELAB® 7000
ELGAPB7060XXM1-230
- Fließrate:Up to 4 l/min
- Bezeichnung:Water purification system, PURELAB® 7000
- Modell:PURELAB® 7000
- Bakterien:<1 CFU/ml
- Speisewasser:Potable tap water
- Max. Arbeitsdruck:6 bar
- Partikel:0,05 µm
- Spezifischer Widerstand bei +25 °C:>15 MΩ.cm
- TOC-Gehalt:<10 ppb
- Gewicht:52 kg
- B×T×H:794×470×820 mm
- VE:1
The PURELAB 7000 series is designed for applications requiring higher flows of Type II water or as a pure water feed system for small distribution loops. The system can supply water from the internal 50 L reservoir at varying flow rates or can feed into much larger vessels as required.
- Small high purity distribution system with variable flow rates
- Auto rinse and simple sanitization procedures maintain the purity of water for the life of the system
- Easy-access doors and castors means system can be located under bench
- Unique RO resin mix to optimise economy, increases yield of purification packs
- Users can easily switch between ultrafiltration or sub-micron filtration for their specific application
Typical applications: AAS, buffer and media preparation, electrophysiology, feed to ultra pure water systems, general chemistry, glassware washing/rinsing, histology, microbiological analysis, RIA/ELISA, sample diluting and reagent preparation, spectrophotometry, water analysis.