ContactUsLinkComponent
Test kits, gold, Spectroquant®, Supelco®
Test kits, gold, Spectroquant®, Supelco®
Catalog # 1.14821.0002
Supplier:  Merck
CAS Number:  
undefined
Test kits, gold, Spectroquant®, Supelco®
Catalog # 1.14821.0002
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Rhodamine B
  • Pk:
    75 Tests
  • Description:
    Gold test
  • M-Clarity quality level:
    MQ100
  • For unit:
    Prove, Nova 60
  • Range:
    0,5 - 12,0 mg/l Au
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.