Order Entry
Switzerland
ContactUsLinkComponent
Anti-TNFA Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: MP6-XT22]
Anti-TNFA Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: MP6-XT22]
Catalog # BLEG506339
Supplier:  Biolegend
Anti-TNFA Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: MP6-XT22]
Catalog # BLEG506339
Supplier:  Biolegend

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Alpha
  • Antigen symbol:
    TNFA
  • Clonality:
    Monoclonal
  • Clone:
    MP6-XT22
  • Conjugation:
    Brilliant Violet® 510
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Gene ID:
    21926
  • Antigen synonyms:
    tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
  • Storage temperature:
    The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    Ambient RT
  • Immunogen:
    E. coli -expressed, recombinant mouse TNF-alpha
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510® under optimal conditions. The solution is free of unconjugated Brilliant Violet 510® and unconjugated antibody.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse