Anti-SIGLECH Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 551]
Supplier: Biolegend
Certificates
About this item
Type: Primary
Antigen: SIGLECH
Clonality: Monoclonal
Clone: 551
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Specifications
- Catalog No:
- BLEG129611
- BLEG129612
- Antigen symbol:
- SIGLECH
- SIGLECH
- Antigen name:
- Sialic Acid Binding Ig-like Lectin H
- Sialic Acid Binding Ig-like Lectin H
- Antigen synonyms:
- Siglec-H|6430529G09Rik
- Siglec-H|6430529G09Rik
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 551
- 551
- Reactivity:
- Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Host:
- Rat
- Rat
- Gene ID:
- 233274
- 233274
- Isotype:
- IgG1
- IgG1
- Immunogen:
- Siglec-H-Fc recombinant fusion protein
- Siglec-H-Fc recombinant fusion protein
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together