Order Entry
Switzerland
ContactUsLinkComponent
Anti-KLRB1C Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: PK136]
Anti-KLRB1C Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: PK136]
Catalog # BLEG108740
Supplier:  Biolegend
Anti-KLRB1C Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: PK136]
Catalog # BLEG108740
Supplier:  Biolegend

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Killer Cell Lectin-Like Receptor Subfamily B, Member 1
  • Antigen symbol:
    KLRB1C
  • Clonality:
    Monoclonal
  • Clone:
    PK136
  • Conjugation:
    Brilliant Violet® 605
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Gene ID:
    17059
  • Antigen synonyms:
    NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • Storage temperature:
    The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    Ambient RT
  • Immunogen:
    NK-1+ cells from mouse spleen and bone marrow
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605® under optimal conditions. The solution is free of unconjugated Brilliant Violet 605® and unconjugated antibody.
  • Size:
    500 µL
  • Pk:
    500 µl

Specifications

About this item

Type: Primary
Antigen: KLRB1C
Clonality: Monoclonal
Clone: PK136
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse