Anti-IL10 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: JES5-16E3]
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: JES5-16E3
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
- Catalog No:
- BLEG505025
- BLEG505026
- Antigen symbol:
- IL10
- IL10
- Antigen name:
- Interleukin 10
- Interleukin 10
- Antigen synonyms:
- IL10A|IL-10|GVHDS|TGIF|CSIF
- IL10A|IL-10|GVHDS|TGIF|CSIF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy7®
- PE (Phycoerythrin)/Cy7®
- Clone:
- JES5-16E3
- JES5-16E3
- Reactivity:
- Mouse
- Baboon, Capuchin monkey, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Host:
- Rat
- Rat
- Gene ID:
- 16153
- 16153
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Immunogen:
- E. coli -expressed, recombinant mouse IL-10
- E. coli -expressed, recombinant mouse IL-10
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together