Mouse IgG2b kappa Isotype Control [clone: MPC-11] (Brilliant Violet® 711)
BLEG400354
- Antibody type:Control
- Antigen symbol:IgG2b kappa
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:Brilliant Violet® 711
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0,2 mg/ml
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711® under optimal conditions. The solution is free of unconjugated Brilliant Violet 711® and unconjugated antibody.
- Size:100 Tests
- Pk:100
Type: Control
Antigen: IgG2b κ
Clonality: Monoclonal
Clone: MPC-11
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Mouse