Order Entry
Switzerland
ContactUsLinkComponent
Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 785)
Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 785)
  BLEG400273
 :  Biolegend
Mouse IgG2a kappa Isotype Control [clone: MOPC-173] (Brilliant Violet® 785)
  BLEG400273
 :  Biolegend

 

  • Antibody type:
    Control
  • Antigen symbol:
    IgG2a
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-173
  • Conjugation:
    Brilliant Violet® 785
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    Ambient RT
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785® under optimal conditions. The solution is free of unconjugated Brilliant Violet 785® and unconjugated antibody.
  • Size:
    25 tests
  • Pk:
    25 Tests

 

 

Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse