Rat IgG1 kappa Isotype Control [clone: RTK2071] (Brilliant Violet® 711)
Catalog # BLEG400441
Supplier: Biolegend
Specifications
- Antibody type:Control
- Antigen symbol:IgG1 kappa
- Clonality:Monoclonal
- Clone:RTK2071
- Conjugation:Brilliant Violet® 711
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Epitope:kappa-chain
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0,2 mg/ml
- Shipping temperature:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711® under optimal conditions. The solution is free of unconjugated Brilliant Violet 711® and unconjugated antibody.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Control
Antigen: IgG1 kappa
Clonality: Monoclonal
Clone: RTK2071
Conjugation: Brilliant Violet® 711
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Rat