Rat IgG1 kappa Isotype Control [clone: RTK2071] (Brilliant Violet® 510)
Catalog # BLEG400435
Supplier: Biolegend
Specifications
- Antikörper-Typ:Kontrolle
- Antigen-Symbol:IgG1 kappa
- Klonalität:Monoklonal
- Klon:RTK2071
- Konjugation:Brilliant Violet® 510
- Wirt:Rat
- Isotyp:IgG1 kappa
- Reaktivität:Mouse
- Epitop:kappa-chain
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Lagertemperatur:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Transporttemperatur:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Reinigung:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510® under optimal conditions. The solution is free of unconjugated Brilliant Violet 510® and unconjugated antibody.
- Größe:125 µL
- VE:125 µl
Specifications
About this item
Type: Control
Antigen: IgG1 kappa
Clonality: Monoclonal
Clone: RTK2071
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Rat