Anti-CCP1 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: GB11]
Anbieter: Biolegend
Zertifikate
Über diesen Artikel
Type: Primary
Antigen: CCP1
Clonality: Monoclonal
Clone: GB11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Technische Daten
- Catalog No:
- BLEG515407
- BLEG515408
- Antigen symbol:
- GZMB
- GZMB
- Antigen name:
- Granzyme B
- Granzyme B
- Antigen synonyms:
- Granzyme-2|serine protease B|CTLA-1|Asp-ase
- Granzyme-2|serine protease B|CTLA-1|Asp-ase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PB (Pacific Blue)
- PB (Pacific Blue)
- Clone:
- GB11
- GB11
- Reactivity:
- Mouse
- Baboon, Pig, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3002, 14939, 171528
- 3002, 14939, 171528
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Pacific Blue® under optimal conditions. The solution is free of unconjugated Pacific Blue®.
- The antibody was purified by affinity chromatography and conjugated with Pacific Blue® under optimal conditions. The solution is free of unconjugated Pacific Blue®.
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
Technische Daten
Frequently Bought Together