Anti-FCAR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: A59]
Catalog # BLEG354105
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgA receptor
- Antigen symbol:FCAR
- Clonality:Monoclonal
- Clone:A59
- Conjugation:APC (Allophycocyanin)
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:2204
- Antigen synonyms:FcalphaRI|CTB-61M7.2|CD89
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:Ag8.653 myeloma cells
- Purification:The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Size:25 Tests
- Pk:25 Tests
Specifications
About this item
Type: Primary
Antigen: FCAR
Clonality: Monoclonal
Clone: A59
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Cynomolgus, Rhesus