Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: IT2.2]
Codice catalogo BLEG305431
Nome del fornitore: Biolegend
Specifiche
- Antibody type:Primary
- Antigen name:CD86 molecule
- Antigen symbol:CD86
- Clonality:Monoclonal
- Clone:IT2.2
- Conjugation:Brilliant Violet® 510
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:942
- Antigen synonyms:CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510® under optimal conditions. The solution is free of unconjugated Brilliant Violet 510® and unconjugated antibody.
- Size:25 tests
- Pk:25
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: IT2.2
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: