Anti-CEACAM8 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: G10F5]
Catalog # BLEG305111
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:Carcinoembryonic antigen-related cell adhesion molecule 8
- Antigen symbol:CEACAM8
- Clonality:Monoclonal
- Clone:G10F5
- Conjugation:PB (Pacific Blue)
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:1088
- Antigen synonyms:CEACAM8|CD66b|NCA-95|CGM6|CD67
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Pacific Blue® under optimal conditions. The solution is free of unconjugated Pacific Blue®.
- Size:25 tests
- Pk:25 Tests
Specifications
About this item
Type: Primary
Antigen: CEACAM8
Clonality: Monoclonal
Clone: G10F5
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: