Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD62P Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMP-1]
Anti-CD62P Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMP-1]
Catalog # BLEG148306
Supplier:  Biolegend
Anti-CD62P Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMP-1]
Catalog # BLEG148306
Supplier:  Biolegend

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    P-selectin
  • Antigen symbol:
    CD62P
  • Clonality:
    Monoclonal
  • Clone:
    RMP-1
  • Conjugation:
    PE (Phycoerythrin)
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Rhesus monkey,
    Pigtailed macaque,
    Chimpanzee,
    Human
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    20344, 25651
  • Antigen synonyms:
    GMP-140|GMP140|platelet activation-dependent granule membrane protein (PADGEM)
  • Storage temperature:
    The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    Ambient RT
  • Immunogen:
    Thrombin activated rat platelets
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CD62P
Clonality: Monoclonal
Clone: RMP-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: