Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-B3GAT1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: HNK-1]
 :  Biolegend
 
Anti-B3GAT1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: HNK-1]
 :  Biolegend

 

  • Catalog No:
  • BLEG359603
  • BLEG359604
  • Antigen symbol:
  • B3GAT1
  • B3GAT1
  • Antigen name:
  • beta-1,3-glucuronyltransferase 1
  • beta-1,3-glucuronyltransferase 1
  • Antigen synonyms:
  • LEU7|GLCATP|CD57|NK1|GLCUATP|NK-1|HNK1
  • LEU7|GLCATP|CD57|NK1|GLCUATP|NK-1|HNK1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • HNK-1
  • HNK-1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 27087
  • 27087
  • Isotype:
  • IgM kappa
  • IgM kappa
  • Immunogen:
  • Membrane extract of human lymphoblastoid cell line HSB-2.
  • Membrane extract of human lymphoblastoid cell line HSB-2.
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC and unconjugated antibody.
  • Storage temperature:
  • The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT

 

Frequently Bought Together