Anti-PTPRC Rat Monoclonal Antibody (Brilliant Violet® 785) [clone: 30-F11]
Catalog # BLEG103149
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:30-F11
- Conjugation:Brilliant Violet® 785
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Rat,Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:19264
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0,2 mg/ml
- Shipping temperature:Ambient RT
- Immunogen:Mouse thymus or spleen
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785® under optimal conditions. The solution is free of unconjugated Brilliant Violet 785® and unconjugated antibody.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: Brilliant Violet® 785
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse