Anti-CD40 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 5C3]
BLEG334328
- Antibody type:Primary
- Antigen name:CD40 Molecule
- Antigen symbol:CD40
- Clonality:Monoclonal
- Clone:5C3
- Conjugation:Alexa Fluor® 700
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rat
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:958
- Antigen synonyms:TNF receptor superfamily member 5|TRAP|AI326936|p50|IMD3|GP39|Tnfrsf5|IGM|T-BAM|Bp50|HIGM1
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
- Size:100 Tests
- Pk:100
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: