Anti-HAVCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)rCP/Cy5.5) [clone: F38-2E2]
Catalog # BLEG345016
Anbieter: Biolegend
Technische Daten
- Antibody type:Primary
- Antigen name:Hepatitis A virus cellular receptor 2
- Antigen symbol:HAVCR2
- Clonality:Monoclonal
- Clone:F38-2E2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:84868
- Antigen synonyms:kidney injury molecule 3|KIM-3|TIMD3|FLJ14428|hepatitis A virus cellular receptor 2|T cell immunoglobulin mucin 3|TIM3|HAVCR2
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:Human Tim-3 fusion protein
- Purification:The antibody was purified by affinity chromatography and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
- Size:100 Tests
- Pk:100 Tests
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: HAVCR2
Clonality: Monoclonal
Clone: F38-2E2
Conjugation: PE (Phycoerythrin)rCP/Cy5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rhesus