Anti-CLEC4C Mouse Monoclonal Antibody () [clone: 201A]
Catalog # BLEG354215-100UG
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:C-type lectin Domain family 4, member C
- Antigen symbol:CLEC4C
- Clonality:Monoclonal
- Clone:201A
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and EDTA.
- Gene ID:170482
- Antigen synonyms:PRO34150|CLECSF11|CD303|BDCA2|HECL|CLECSF7|DLEC
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C.
- Concentration:1,0 mg/ml
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CLEC4C
Clonality: Monoclonal
Clone: 201A
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: