Order Entry
Switzerland
ContactUsLinkComponent
Anti-ATP1B3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LNH-94]
Catalog # BLEG341706
Supplier:  Biolegend
Anti-ATP1B3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LNH-94]
Catalog # BLEG341706
Supplier:  Biolegend

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, Na+/K+ transporting, beta 3 polypeptide
  • Antigen symbol:
    ATP1B3
  • Clonality:
    Monoclonal
  • Clone:
    LNH-94
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Gene ID:
    483
  • Antigen synonyms:
    ATPB-3|CD298
  • Storage temperature:
    The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    Ambient RT
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: ATP1B3
Clonality: Monoclonal
Clone: LNH-94
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human