Anti-CD274 Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 10F.9G2]
Catalog # BLEG124319
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:CD274 molecule
- Antigen symbol:CD274
- Clonality:Monoclonal
- Clone:10F.9G2
- Conjugation:Brilliant Violet® 711
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:60533
- Antigen synonyms:PDL1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0,2 mg/ml
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711® under optimal conditions. The solution is free of unconjugated Brilliant Violet 711® and unconjugated antibody.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 10F.9G2
Conjugation: Brilliant Violet® 711
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse