Anti-CD27 Mouse Monoclonal Antibody () [clone: O323]
BLEG302839
- Antibody type:Primary
- Antigen name:CD27 molecule
- Antigen symbol:CD27
- Clonality:Monoclonal
- Clone:O323
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and EDTA.
- Gene ID:939
- Antigen synonyms:Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C.
- Concentration:1,0 mg/ml
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Baboon, Cynomolgus, Rhesus, Squirrel Monkey, Squirrel Monkey