Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD226 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11A8]
Anti-CD226 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11A8]
Codice catalogo BLEG338312
Nome del fornitore:  Biolegend
Anti-CD226 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11A8]
Codice catalogo BLEG338312
Nome del fornitore:  Biolegend

Specifiche

  • Antibody type:
    Primary
  • Antigen name:
    CD226 molecule
  • Antigen symbol:
    CD226
  • Clonality:
    Monoclonal
  • Clone:
    11A8
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Bovine
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Gene ID:
    10666
  • Antigen synonyms:
    TLiSA1|PTA1|DNAM1|CD226 antigen|Adhesion Glycoprotein|Platelet and T cell Activation Antigen 1|CD226 molecule|T lineage Specific Activation Antigen 1|DNAM 1|DNAX Accessory Molecule 1
  • Storage temperature:
    The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    Ambient RT
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Size:
    100 Tests
  • Pk:
    100

Specifiche

Informazioni su questo articolo

Type: Primary
Antigen: CD226
Clonality: Monoclonal
Clone: 11A8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human