Anti-CCR3 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 5E8]
BLEG310714
- Antikörper-Typ:Primär
- Antigen-Name:Chemokine (C-C motif) receptor 3
- Antigen-Symbol:CCR3
- Klonalität:Monoklonal
- Klon:5E8
- Konjugation:Brilliant Violet® 421
- Wirt:Mouse
- Isotyp:IgG2b kappa
- Reaktivität:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gen-ID:1232
- Antigen-Synonyme:CD193|eotaxin receptor|MIP1-alpha receptor like-2|CC CKR3
- Lagertemperatur:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Transporttemperatur:Ambient RT
- Reinigung:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421® under optimal conditions. The solution is free of unconjugated Brilliant Violet 421® and unconjugated antibody.
- Größe:100 Tests
- VE:100 Tests
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 500000000
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human