Anti-CXCR5 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: L138D7]
Katalognr. BLEG145512
Anbieter: Biolegend
Technische Daten
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) Receptor 5
- Antigen symbol:CXCR5
- Clonality:Monoclonal
- Clone:L138D7
- Conjugation:Brilliant Violet® 421
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:12145
- Antigen synonyms:CD185|MDR15|CXCR-5|Gpcr6|Blr1|CXC-R5
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:mCXCR5-transfected cells
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421® under optimal conditions. The solution is free of unconjugated Brilliant Violet 421® and unconjugated antibody.
- Size:500 µL
- Pk:500 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: L138D7
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype:
Reactivity: