Anti-CXCR5 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: J252D4]
Catalog # BLEG356912
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) Receptor 5
- Antigen symbol:CXCR5
- Clonality:Monoclonal
- Clone:J252D4
- Conjugation:Alexa Fluor® 488
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:643
- Antigen synonyms:CD185|MDR15|CXCR-5|Gpcr6|Blr1|CXC-R5
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:Human CXCR5-transfected cells
- Purification:The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 488 under optimal conditions.
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: J252D4
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: