Anti-PVR Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: TX56]
Catalog # BLEG131511
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:Poliovirus Receptor
- Antigen symbol:PVR
- Clonality:Monoclonal
- Clone:TX56
- Conjugation:PE (Phycoerythrin)/Cy7®
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:52118
- Antigen synonyms:TAGE4|PVS|CD155|NECL5|Necl-5|HVED
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0,2 mg/ml
- Shipping temperature:Ambient RT
- Immunogen:CD155 transfectants
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- Size:25 µg
- Pk:25 µG
Specifications
About this item
Type: Primary
Antigen: PVR
Clonality: Monoclonal
Clone: TX56
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: rat
Isotype: IgG2a kappa
Reactivity: Mouse