Anti-ITGAM Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: M1/70]
Catalog # BLEG101245
Supplier: Biolegend
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha M
- Antigen symbol:ITGAM
- Clonality:Monoclonal
- Clone:M1/70
- Conjugation:Brilliant Violet® 510
- Flow cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:16409
- Antigen synonyms:CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:C57BL/10 splenocytes
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510® under optimal conditions. The solution is free of unconjugated Brilliant Violet 510® and unconjugated antibody.
- Size:125 µL
- Pk:125 µl
Specifications
About this item
Type:
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee, Rabbit