ContactUsLinkComponent
Net dressing for the head
Net dressing for the head
Catalog # CEDE1893
Supplier:  ORKLA CARE
undefined
Net dressing for the head
Catalog # CEDE1893
Supplier:  ORKLA CARE
Specifications
  • Description:
    Large sterile metallised dressing and latex-free tubular net for keeping the dressing in place
  • Pk:
    1
Specifications
About this item
This kit contains a large sterile metallised dressing and latex-free tubular net intended for keeping the dressing in place. The dressing is intended to be used as a mechanical barrier and/or for absorption of exudates from large wounds on the head or other parts of the body, e.g. legs.
  • Dressing: Absorbent layer made of non woven 100% viscose; non adherent layer of PE on both sides of dressing; aluminium metallised on one side
  • Non adherent layer on both sides of the dressing prevents the dressing from sticking to the wound
  • Metallised dressings are sterilised by irradiation
Dressing: 200×400 mm
Tubular net: 70×400 mm (unstretched)