ContactUsLinkComponent
Dimroth condensers with standard ground joints
Dimroth condensers with standard ground joints
Catalog # KAVA442517266
Supplier:  Kavalier
undefined
Dimroth condensers with standard ground joints
Catalog # KAVA442517266
Supplier:  Kavalier
Specifications
  • Cone size:
    29/32 NS
  • Jacket length:
    250 mm
  • Socket size:
    29/32 NS
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For cooling condensate from vapours, with core and jacket.