Temperature loggers, ECOLOG TN2/TN3-P
Supplier: ELPRO-BUCHS
Certificates
About this item
ECOLOG TN2/TN3-P data loggers comes with display and alarm function, used for the measurement of temperature.These loggers suitable for foodstuff applications. It has desired time interval (from every 1 seconds to every 3 hours).
- Thermoplastic ABS housing
- Ring buffer or start/stop buffer with external start option
- Low battery warning
PC software elproLOG ANALYZE for all communication, reprogramming, display, statistics and printout.
Certifications: IP 54 rated protection class.
Specifications
- Catalog No:
- ELPR800429
- ELPR800428
- ELPR800430
- Model:
- ECOLOG TN3-P
- ECOLOG TN2
- ECOLOG TN3-PEx
- Sensor:
- 1 built-in NTC sensor –35...+55 °C and up to 2 or 3 external NTC sensors –50...+140 °C
- 1 built-in NTC sensor –35...+55 °C and up to 2 or 3 external NTC sensors –50...+140 °C
- 1 built-in NTC sensor –35...+55 °C and up to 2 or 3 external NTC sensors –50...+140 °C
- Alarm:
- External alarm on DB15 connector and alarm display (programmable)
- External alarm on DB15 connector and alarm display (programmable)
- External alarm on DB15 connector and alarm display (programmable)
- Display:
- LCD
- LCD
- LCD
- Interface:
- RS232
- RS232
- RS232
- Battery life:
- 2 Years
- 2 Years
- 2 Years
- Recording intervals:
- 1 s - 3 h
- 1 s - 3 h
- 1 s - 3 h
- W×D×H:
- 110×85×35 mm
- 110×85×35 mm
- 110×85×35 mm
- Data memory - No. of readings:
- 64000
- 64000
- 64000
- Batteries:
- One lithium battery, 3,6 V
- One lithium battery, 3,6 V
- One lithium battery, 3,6 V
Specifications
Probes
Product Information
Frequently Bought Together