ContactUsLinkComponent
Serological pipettes, Shortie, Nunc™
Serological pipettes, Shortie, Nunc™
Catalog # 612-4741
undefined
Serological pipettes, Shortie, Nunc™
Catalog # 612-4741
Specifications
  • Packed:
    Individually wrapped; 50/bag; 200/case
  • Sterile:
    +
  • Capacity:
    10 ml
  • Seq:
    02
  • Pk:
    200
Specifications
About this item
PS, transparent, graduated and sterile. These sleek pipettes, made of ultra-clear virgin PS, are ideal for easy manoeuvring under the hood.
  • Disposable, plugged
  • Certified non pyrogenic
  • Bold clear graduations with easy to read scale markings
  • Non drip tip for pipette precision
  • Sterility assurance level SAL 10⁻⁶
Packaging: Individually wrapped in paper/plastic peel packaging.