Gel drying frame kit
Supplier: Serva
Certificates
About this item
Gel drying frame kit suitable for drying mini vertical slab gels up to 100×100 mm.
Delivery information: Available as frames only or as complete kit including two frames and 200 cellophane sheets.
Frequently Bought Together