ContactUsLinkComponent
Mini spore discs
Mini spore discs
Catalog # NAMSDN18-06
Supplier:  CROSSTEX
Mini spore discs
Catalog # NAMSDN18-06
Supplier:  CROSSTEX
Specifications
  • Ø:
    3 mm
  • Description:
    Bacillus atrophaeus cell line 9372: mini spore disc 106 (24 month shelf life) / sterilisation process: dry heat
  • Pk:
    100
Specifications
About this item
Inoculated carrier mini spore discs for monitoring EO and dry heat processes.
  • An inoculated carrier, 3 mm disc, of Bacillus atrophaeus (cell Line 9372)
  • Primary packaging in bulk
The mini spore discs are designed to be placed directly into a device and utilised to monitor EO and dry heat sterilisation process efficacy.
The mini spore discs are labelled for industrial use only.
Certifications: ISO 11138-1 Sterilisation of health care products – Biological indicators – Part 1: General requirements.
ISO 11138-2 Sterilisation of health care products – Biological indicators – Part 2: Biological indicators for ethylene oxide sterilisation processes.