Order Entry
Switzerland
ContactUsLinkComponent
Silver nitrate standard solution (1 mL = 1 mg Chloride), BAKER ANALYZED®, J.T.Baker®
Irritant
Environment
Silver nitrate standard solution (1 mL = 1 mg Chloride), BAKER ANALYZED®, J.T.Baker®
Catalog # 5652-03
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Environment
Silver nitrate standard solution (1 mL = 1 mg Chloride), BAKER ANALYZED®, J.T.Baker®
Catalog # 5652-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Titer, mg/mL, For Information Only:
    3.0 - 3.5
  • pH at 25°C:
    3.0 - 3.5
  • Sulfate (SO₄):
    ≤ 1 ppm

Specification Test Results

About this item

 :
 :169.87 g/mol
 :GHS07
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414