Order Entry
Switzerland
ContactUsLinkComponent
Ammonium metavanadate ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Toxic
Ammonium metavanadate ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0715-04
Supplier:  Avantor
CAS Number:  
undefined
Toxic
Ammonium metavanadate ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0715-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    125 g
  • Seq:
    3445

Specifications

About this item

 :
 :116.98 g/mol
Merck Index:13,00572
Melting Pt:200 °C
Density:2.33 g/cm³ (20 °C)
EINECS:232-261-3
UN:2859
ADR:6.1,II
 :GHS06
CAS Number:7803-55-6
Formula:NH₄VO₃
MDL Number:MFCD00011430