


Imidazole ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Merck Index:13,04935
Boiling Pt:255 °C (1013 hPa)
Melting Pt:89…90 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:135 °C
Storage Temperature:Ambient
UN:3263
ADR:8,III
:68.08 g/mol
EINECS:206-019-2
:GHS05
:GHS08
:GHS07
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (C₃H₄N₂):≥99.0 %
- Trace Impurities - Iron (Fe):≤0.001 %
- pH of 5% Solution at 25°C:9.5 - 11.0
- Residue after Ignition:≤0.10 %
- Water (H₂O):≤0.2 %
- DNase Activity, For Information Only:Passes Test
- Protease Activity, For Information Only:Passes Test
- RNase Activity, For Information Only:Passes Test
Specification Test Results
Frequently Bought Together