Order Entry
Switzerland
ContactUsLinkComponent
Sodium iodide ≥99.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium iodide ≥99.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3748-07
Supplier:  Avantor
CAS Number:  
undefined
Sodium iodide ≥99.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3748-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.5%
  • For Laboratory,Research,or Manufacturing Use:
    6.0 - 9.0
  • ACS - Assay (NaI):
    ≥99.5 %
  • ACS - pH of 5% Solution at 25°C:
    6.0 - 9.0
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Chloride and Bromide (as Cl):
    ≤0.01 %
  • ACS - Iodate (IO₃):
    ≤3 ppm
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Barium (Ba):
    ≤0.002 %
  • ACS - Heavy Metals:
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Calcium (Ca):
    ≤0.002 %
  • ACS - Magnesium (Mg):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.01 %

Specification Test Results

About this item

 :
 :149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532