Cyclohexanone ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Glass bottle, amber, narrow mouth, 4 L
Supplier: Avantor
Cyclohexanone ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Catalog # 9210-03
Supplier: Avantor
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Lot analysis on label.
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155.6 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0.9478 g/cm³ (20 °C)
- Flash Pt:43 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (C₆H₁₀O) (by GC):≥ 99.0 %
- Color (APHA):≤ 10
- Density (g/mL) at 25°C:0.940 - 0.943
- Residue after Evaporation:≤ 0.0500 %
- Acidity (meq/g):≤ 0.003
- Water (H₂O)(by Karl Fischer titrn):≤ 0.1 %
- Identification (by IR):Passes Test
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



