


Methanol absolute, anhydrous ≥99.8% (by GC), AR® ACS, Macron Fine Chemicals™
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:11 °C (closed cup)
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS02
:GHS06
:GHS08
- For Laboratory,Research,or Manufacturing Use:≥99.8%
- ACS - Assay (CH₃OH) (by GC):≥99.8 %
- Appearance (clear, colorless liquid):Passes Test
- Carbonyl Compounds - Acetaldehyde:≤0.001 %
- Carbonyl Compounds - Acetone:≤0.001 %
- Carbonyl Compounds - Formaldehyde:≤0.001 %
- Color (APHA):≤10
- Heavy Metals (as Pb):≤0.0001 %
- Identification:Passes Test
- Residue after Evaporation:≤0.001 %
- Solubility in H₂O:Passes Test
- Substances Darkened by H₂SO₄:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Acid (meq/g):≤0.0003
- Titrable Base (meq/g):≤0.0002
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
Frequently Bought Together