


Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:Ambient
:200-659-6
:1230
:3,II
:Management of Change (MOC) category = N
:11 °C (closed cup)
:01-2119433307-44
:GHS02
:GHS06
:GHS08
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Assay (CH₃OH) (by GC):≥ 99.8 %
- Substances Darkened by H₂SO₄:Passes Test
- Substances Reducing KMnO₄:Passes Test
- Solubility in H₂O:Passes Test
- Color (APHA):≤ 10
- Water (by KF, coulometric):≤ 0.1 %
- Residue after Evaporation:≤ 0.001 %
- Carbonyl Compounds - Acetaldehyde:≤ 0.001 %
- Carbonyl Compounds - Acetone:≤ 0.001 %
- Carbonyl Compounds - Formaldehyde:≤ 0.001 %
- Titrable Acid (µeq/g):≤ 0.3
- Titrable Base (µeq/g):≤ 0.2
- Heavy Metals (as Pb):≤ 0.5 ppm
- Trace Impurities - Copper (Cu):≤ 0.10 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Trace Impurities - Nickel (Ni):≤ 0.1 ppm
- Date Received: __ __ /__ __ /__ __:—
- Date Opened: __ __ /__ __ /__ __:—
- Meets Reagent Specifications for testing USP/NF monographs:—
Frequently Bought Together