
Citric acid monohydrate 99.0-102.0% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:5949-29-1
:
:C₆H₈O₇.H₂O
:MFCD00149972
:210.14 g/mol
:13,02350
:135…152 °C
:1.54 g/cm³
:173.9 °C (closed cup)
:201-069-1
:GHS07
:Ambient
- Meets ACS Reagent Chemical Requirements:99.0 - 102.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- Assay (HOC(COOH)(CH₂COOH)₂H₂O) (by acidimetry):99.0 - 102.0 %
- Insoluble Matter:≤ 0.005 %
- Residue on Ignition:≤ 0.02 %
- Chloride (Cl):≤ 0.001 %
- Oxalate (C₂O₄):Passes Test
- Phosphate (PO₄):≤ 0.001 %
- Sulfur Compounds (as SO₄):≤ 0.002 %
- Substances Carbonizable by Hot H₂SO₄:Passes Test
- Trace Impurities - Iron (Fe):≤ 2.0 ppm
- Trace Impurities - Lead (Pb):≤ 2.000 ppm
Frequently Bought Together