Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bottle weights

Bottle weights

Flexible lead bottle cuff with durable PVC coating, inner Ø 53 mm, 59 mm, 240 g
Catalog # 214-1911
Supplier:   KLEINFELD
Bottle weights
Catalog # 214-1911
Supplier:  KLEINFELD
Bottle weights

Flexible lead bottle cuff with durable PVC coating, inner Ø 53 mm, 59 mm, 240 g

Price
SFr. 103.00
Log in for availability
1 * 1 ST
Product Details & Documents

Lead with PVC coating. These cuffs are flexible. They can be opened, closed or adapted to the size of the bottle.

Specifications
  • Weight:
    240 g
  • Description:
    Flexible lead bottle cuff with durable PVC coating, inner Ø 53 mm
  • Seq:
    01
  • Container max. Ø:
    59 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.