ContactUsLinkComponent
Hybrid multi-parameter meter, handheld, SL1000 PPA
 :  Hach
undefined
Hybrid multi-parameter meter, handheld, SL1000 PPA
 :  Hach
  • Catalog No:
  • 705-0942
  • 705-0941
  • Model:
  • SL1000
  • SL1000
  • Temperature compensation:
  • Off, automatic and manual (for IntelliCal™ probes; parameter dependent)
  • Off, automatic and manual (for IntelliCal™ probes; parameter dependent)
  • Power supply:
  • Lithium ion rechargeable battery or external charger (100 - 240 V, 50/60 Hz)
  • Lithium ion rechargeable battery or external charger (100 - 240 V, 50/60 Hz)
  • Compliance:
  • IP 64 with caps installed for the meter probe connector ports or with the probes connected to the instrument.
  • IP 64 with caps installed for the meter probe connector ports or with the probes connected to the instrument.
  • Data logging - log function:
  • Automatic in Press to Read measurement mode; manual in Continuous measurement mode
  • Automatic in Press to Read measurement mode; manual in Continuous measurement mode
  • Data logging capacity:
  • 1000 measured values (result, date, time, site ID, user ID)
  • 1000 measured values (result, date, time, site ID, user ID)
  • Connections:
  • 2× M12 connectors for IntelliCal™ probes, 4× Chemkey slots, 2,5 mm Jack for external power supply
  • 2× M12 connectors for IntelliCal™ probes, 4× Chemkey slots, 2,5 mm Jack for external power supply
  • Outputs:
  • Mini USB
  • Mini USB
  • W×D×H:
  • 131×59×258 mm
  • 131×59×258 mm
  • Weight:
  • 1,2 kg
  • 1,2 kg
Reagents for disinfection control
Frequently Bought Together