Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Methanol : Water (30:70) % (v/v), LiChrosolv®, Supelco®
  4.80508.9030
 :  Merck
 :  
 
 
 
Methanol : Water (30:70) % (v/v), LiChrosolv®, Supelco®
  4.80508.9030
 :  Merck
 :  

 

  • Pack type:
    Stainless steel barrel, returnable
  • Pk:
    30 L
  • Seq:
    5352

 

 

  • Density:
    0.95 g/cm³ (20 °C)
  • Flash point:
    35 °C
  • Storage class:
    3 Flammable liquids
  • WGK:
    WGK 1 slightly water endangering
  • R Phrase:
    R 10-23/24/25-39/23/24/25
  • S Phrase:
    S 36/37-45

 

 

 :
 :3,III
 :M-Clarity™ quality level = MQ100
 :1987
 :GHS02
 :GHS08
 :GHS07
 :Ambient
 :67-56-1
 :CH₄O/H₂O
 :MFCD00004595