Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Thorin indicator for sulfate titration, Supelco®
  1.08294.0005
 :  Merck
 :  
undefined
 
 
Thorin indicator for sulfate titration, Supelco®
  1.08294.0005
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    576.3 g/mol
  • Bulk Density:
    700 kg/m3
  • pH:
    6.1 (10 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials
  • Storage:
    +5...+30 °C
  • Absorption maximum λmax. (Propan-2-ol):
    480 - 490 nm
  • Suitability as indicator:
    Passes test
  • Identity:
    Passes test
  • For:
    Sulfate titration

 

 

 :
 :576.30 g/mol
 :6.1,II
 :M-Clarity™ quality level = MQ200
 :222-993-1
 :3465
 :GHS06
 :GHS09
 :Ambient
 :3688-92-4
 :C₁₆H₁₁AsN₂Na₂O₁₀S₂
 :MFCD00003888