
PSS (phosphoric acid, sulphuric acid), Spectrosolv® acid blank, for the ACIDORAPID® mineralizer, Supelco®
Catalog # 1.07916.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.78 g/cm3 (20 °C)
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:196.08 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3264
:GHS05
Storage Temperature:Ambient
Formula:H₅O₈PS