Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Hydrochloric acid 36.5 - 38.0% NF, FCC, ACS, Macron Fine Chemicals™
 
undefined
 
 
Hydrochloric acid 36.5 - 38.0% NF, FCC, ACS, Macron Fine Chemicals™
 

 

  • Meets N.F. Requirements:
    36.5 - 38.0 %
  • Food GMP Manufactured Product:
    ≤ 3 ppm
  • Store below 25°C:
    ≤ 0.010 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤ 0.005 %
  • Bulk Food Chemical:
    ≤ 10
  • Bulk Pharmaceutical Chemical:
    ≤ 5 ppm
  • ACS - Assay (HCl) (ACS):
    36.5 - 38.0 %
  • ACS - Ammonium (NH₄):
    ≤ 3 ppm
  • ACS - Arsenic (As):
    ≤ 0.010 ppm
  • ACS - Bromide (Br):
    ≤ 0.005 %
  • ACS - Color (APHA):
    ≤ 10
  • ACS - Extractable Organic Substances:
    ≤ 5 ppm
  • ACS - Free Chlorine (as Cl):
    ≤ 1 ppm
  • ACS - Residue after Ignition:
    ≤ 5 ppm
  • ACS - Sulfate (SO₄):
    ≤ 1 ppm
  • ACS - Sulfite (SO₃):
    ≤ 1 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 1 ppm
  • Trace Impurities - ACS - Iron (Fe):
    ≤ 0.200 ppm
  • NF - Assay (HCl)(NF):
    36.5 - 38.0 %
  • NF - Bromide or Iodide:
    Passes Test
  • NF - Free Bromine or Chlorine:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Residue on Ignition:
    ≤ 0.008 %
  • NF - Sulfate (NF)(SO₄):
    Passes Test
  • NF - Sulfite (NF)(SO₃):
    Passes Test
  • FCC - Assay (HCl)(FCC):
    36.5 - 38.0 %
  • FCC - Baume Degrees:
    22.7 - 23.4
  • FCC - Color (APHA):
    Passes Test
  • FCC - Identification:
    Passes Test
  • FCC - Iron (Fe):
    ≤ 0.2 ppm
  • FCC - Lead (Pb) (mg/kg):
    ≤ 1
  • FCC - Mercury (Hg)(mg/kg):
    ≤ 0.10
  • FCC - Nonvolatile Residue:
    ≤ 0.5 %
  • Total Organic Compounds - Non-Fluorine Containing (mg/kg):
    ≤ 5
  • Total Organic Compounds - Benzene (mg/kg):
    ≤ 0.05
  • Total Organic Compounds - Fluorinated Organic Compounds(mg/kg):
    ≤ 25
  • Total Organic Compounds - Oxidizing Substances:
    ≤ 0.003 %
  • Reducing Substances:
    ≤ 0.007 %
  • Specific Gravity:
    1.1856 - 1.1924
  • Sulfate (SO₄):
    ≤ 0.5 %
  • Appearance:
    Passes Test

 

Frequently Bought Together