Smoking receptacle, Cease Fire®
Original Smoker’s Cease Fire® cigarette butt receptacle, Grey, 15 L, 419×978 mm
Catalog # 119-0283
Supplier: JUSTRITE MANUFACTURING
Jump to:
Product Details & Documents
The original smoker’s Cease Fire® cigarette butt receptacle’s self-extinguishing design safely collects unsightly cigarette litter and reduces the risk of fire. Unique rain bonnet opening keeps rain out and discourages other trash. Has a big four gallon galvanised steel pail inside. 'Drip-lip' internal construction prevents tar condensation from escaping, keeping exterior clean from dark, messy drip marks.
- Made of flame-retardant, sturdy polyethylene
- Tie-down notches offer security and greater stability under severe weather conditions
- Fire tested and approved by FM Global
Specifications
- Ø×H:419×978 mm
- Capacity:15 L
- Colour:Grey
- Description:Original Smoker’s Cease Fire® cigarette butt receptacle
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



